Learn More
Medchemexpress LLC Sertraline hydrochloride | 79559-97-0 | 99.9% | 342.69 | 50 MG
SDP
Catalog No. 5000364488
Encompass_Preferred
Sertraline hydrochloride is an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. It is researched for a number of diseases, such as major depressive disorder and obsessive-compulsive disorder.
  • Exhibits spermicidal activity against human spermatozoa.
  • Inhibits microglial activation and inflammatory mediator production in vitro.
  • Reduces immobility time in the forced swim test in female animals.
  • Does not alter spontaneous locomotor activity across various genotypes.
  • Shows anti-depression effects in chronic unpredictable mild stress models.
Catalog No. 50-003-64488 Supplier Medchemexpress LLC Supplier No. 50MGHYB0176A
Sertraline hydrochloride is an antidepressant of the selective serotonin reuptake inhibitor (SSRI) class. It is researched for a number of diseases, such as major depressive disorder and obsessive-compulsive disorder.
  • Exhibits spermicidal activity against human spermatozoa.
  • Inhibits microglial activation and inflammatory mediator production in vitro.
  • Reduces immobility time in the forced swim test in female animals.
  • Does not alter spontaneous locomotor activity across various genotypes.
  • Shows anti-depression effects in chronic unpredictable mild stress models.