Learn More
Medchemexpress LLC Sertraline hydrochloride | 79559-97-0 | MFCD00895772 | 99.9% | 1 ML
SDP
Catalog No. 5000406314
Encompass_Preferred
Sertraline hydrochloride is an antidepressant classified as a selective serotonin reuptake inhibitor (SSRI). It is currently being researched for the treatment of various conditions, including major depressive disorder and obsessive-compulsive disorder.
  • Shows spermicidal activity against human spermatozoa.
  • Inhibits microglial activation and inflammatory mediator production.
  • Produces anti-depression effects in CUMS-exposed mice.
  • Reduced immobility time during forced swim test in female animals.
Catalog No. 50-004-06314 Supplier Medchemexpress LLC Supplier No. HYB0176A10MM/1ML
Sertraline hydrochloride is an antidepressant classified as a selective serotonin reuptake inhibitor (SSRI). It is currently being researched for the treatment of various conditions, including major depressive disorder and obsessive-compulsive disorder.
  • Shows spermicidal activity against human spermatozoa.
  • Inhibits microglial activation and inflammatory mediator production.
  • Produces anti-depression effects in CUMS-exposed mice.
  • Reduced immobility time during forced swim test in female animals.