Learn More
Abcam Sertraline hydrochloride, Serotonin reuptake inhibitor, 50MG
Catalog No. 502706934
Encompass_Preferred
MW 306.2 Da, Purity >99%. Selective serotonin reuptake inhibitor (Ki = 2.0 μM). Shows antidepressant and antiproliferative activity. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6934 Supplier Abcam Supplier No. AB14106850MG
MW 306.2 Da, Purity >99%. Selective serotonin reuptake inhibitor (Ki = 2.0 μM). Shows antidepressant and antiproliferative activity. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement