Learn More
Cayman Chemical Sertraline (hydrochloride), Purity: ≥95%, CAS: 79559-97-0, 50 mg
SDP
Catalog No. 502972212
Encompass_Preferred
A selective serotonin reuptake inhibitor; inhibits monoamine uptake by SERT (IC50 = 70 nM); selective for SERT over the norepinephrine and dopamine transporters (IC50s = 520 and 720 nM, respectively); reduces immobility in the forced swim test in mice at 3.2-56 mg/kg, s.c.; prevents acid sphingomyelinase activation and subsequent ceramide release induced by pp-VSV-SARS-CoV-2 infection in Vero cells at 5 µM
Catalog No. 50-297-2212 Supplier Cayman Chemical Supplier No. 1483950 MG
A selective serotonin reuptake inhibitor; inhibits monoamine uptake by SERT (IC50 = 70 nM); selective for SERT over the norepinephrine and dopamine transporters (IC50s = 520 and 720 nM, respectively); reduces immobility in the forced swim test in mice at 3.2-56 mg/kg, s.c.; prevents acid sphingomyelinase activation and subsequent ceramide release induced by pp-VSV-SARS-CoV-2 infection in Vero cells at 5 µM