Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Serum albumin/ALB protein, human (HEK293, His) | >95.0% | 200UG
SDP

Catalog No. 5000174046
Encompass_Preferred

Recombinant human serum albumin produced in HEK293 cells with a C-terminal His tag, supplied as a purified protein (200 μg) for research use. It offers high purity and a defined molecular weight, suitable for binding studies, assay development, and as a carrier or stabilizer. Follow the supplied datasheet for reconstitution and handling instructions.

  • His-tagged human serum albumin expressed in HEK293 cells
  • Purity >95% as determined by reducing SDS-PAGE
  • Molecular weight 67-80 kDa
  • Supplied as 200 μg of purified protein
  • Not recommended to reconstitute below 100 μg/mL in ddH2O
  • After reconstitution stable at 4°C for 1 week or -20°C for longer
  • Freeze aliquots at -20°C or -80°C for extended storage

Catalog No. 50-001-74046 Supplier Medchemexpress LLC Supplier No. HYP704297200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human serum albumin produced in HEK293 cells with a C-terminal His tag, supplied as a purified protein (200 μg) for research use. It offers high purity and a defined molecular weight, suitable for binding studies, assay development, and as a carrier or stabilizer. Follow the supplied datasheet for reconstitution and handling instructions.

  • His-tagged human serum albumin expressed in HEK293 cells
  • Purity >95% as determined by reducing SDS-PAGE
  • Molecular weight 67-80 kDa
  • Supplied as 200 μg of purified protein
  • Not recommended to reconstitute below 100 μg/mL in ddH2O
  • After reconstitution stable at 4°C for 1 week or -20°C for longer
  • Freeze aliquots at -20°C or -80°C for extended storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.