Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Serum Albumin/ALB C 10ug
SDP

Catalog No. 5000195920
Encompass_Preferred

Recombinant serum albumin (ALB) derived from cynomolgus, produced in Pichia pastoris and supplied with an N-terminal His tag. It is intended as a carrier and control protein for biochemical and cell-based assays, exhibiting broad binding to ions, fatty acids, hormones, bilirubin, and various small molecules. Supplied in small aliquot sizes for assay development and quality control.

  • Recombinant protein produced in P. pastoris with an N-terminal His tag.
  • Binds water, metal ions, fatty acids, hormones, bilirubin, and small molecules.
  • Sequence corresponds to residues D25-A608 with an approximate molecular weight of 68.0 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Stable when stored at -20°C; aliquot and freeze at -80°C for extended storage.
  • Available in small pack sizes suitable for assay controls and binding studies.

Catalog No. 50-001-95920 Supplier Medchemexpress LLC Supplier No. HYP7171110UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant serum albumin (ALB) derived from cynomolgus, produced in Pichia pastoris and supplied with an N-terminal His tag. It is intended as a carrier and control protein for biochemical and cell-based assays, exhibiting broad binding to ions, fatty acids, hormones, bilirubin, and various small molecules. Supplied in small aliquot sizes for assay development and quality control.

  • Recombinant protein produced in P. pastoris with an N-terminal His tag.
  • Binds water, metal ions, fatty acids, hormones, bilirubin, and small molecules.
  • Sequence corresponds to residues D25-A608 with an approximate molecular weight of 68.0 kDa.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Stable when stored at -20°C; aliquot and freeze at -80°C for extended storage.
  • Available in small pack sizes suitable for assay controls and binding studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.