Learn More
Medchemexpress LLC Serum Albumin/ALB C 50ug | 50ug
SDP
Catalog No. 5000195921
Encompass_Preferred
Serum albumin (ALB) protein binds a variety of substances including water calcium sodium potassium fatty acids hormones bilirubin and drugs It regulates blood osmolality and transports zinc calcium and magnesium Serum Albumin/ALB Protein Cynomolgus (P pastoris His) is the recombinant cynomolgus-derived Serum Albumin/ALB protein expressed by P pastoris with N-His labeled tag
Catalog No. 50-001-95921 Supplier Medchemexpress LLC Supplier No. HYP7171150UG
Serum albumin (ALB) protein binds a variety of substances including water calcium sodium potassium fatty acids hormones bilirubin and drugs It regulates blood osmolality and transports zinc calcium and magnesium Serum Albumin/ALB Protein Cynomolgus (P pastoris His) is the recombinant cynomolgus-derived Serum Albumin/ALB protein expressed by P pastoris with N-His labeled tag