Learn More
Medchemexpress LLC Serum Albumin/ALB H 20ug | 20ug
SDP
Catalog No. 5000187804
Encompass_Preferred
The serum albumin/ALB protein has multiple binding abilities and can bind water Ca(2 ) Na( ) K( ) fatty acids hormones bilirubin and drugs Its main effects include regulating blood colloid osmotic pressure to achieve homeostasis Serum Albumin/ALB Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Serum Albumin/ALB protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87804 Supplier Medchemexpress LLC Supplier No. HYP7807720UG
The serum albumin/ALB protein has multiple binding abilities and can bind water Ca(2 ) Na( ) K( ) fatty acids hormones bilirubin and drugs Its main effects include regulating blood colloid osmotic pressure to achieve homeostasis Serum Albumin/ALB Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Serum Albumin/ALB protein expressed by HEK293 with C-Avi C-His labeled tag