Learn More
Medchemexpress LLC Serum Albumin/ALB H 50ug | 50ug
SDP
Catalog No. 5000194046
Encompass_Preferred
The serum albumin/ALB protein has multiple binding abilities and can bind water Ca(2 ) Na( ) K( ) fatty acids hormones bilirubin and drugs Its main effects include regulating blood colloid osmotic pressure to achieve homeostasis Serum Albumin/ALB Protein Human (HEK293 His-Avi) is the recombinant human-derived Serum Albumin/ALB protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-94046 Supplier Medchemexpress LLC Supplier No. HYP7839350UG
The serum albumin/ALB protein has multiple binding abilities and can bind water Ca(2 ) Na( ) K( ) fatty acids hormones bilirubin and drugs Its main effects include regulating blood colloid osmotic pressure to achieve homeostasis Serum Albumin/ALB Protein Human (HEK293 His-Avi) is the recombinant human-derived Serum Albumin/ALB protein expressed by HEK293 with C-Avi C-His labeled tag