Learn More
Medchemexpress LLC Serum Albumin/ALB M 100ug | 100ug
SDP
Catalog No. 5000193746
Encompass_Preferred
Serum albumin/ALB protein binds to various substances including water calcium sodium potassium fatty acids hormones bilirubin and drugs Its main function is to regulate blood osmotic pressure and transport zinc calcium and magnesium Serum Albumin/ALB Protein Mouse (HEK293 His) is the recombinant mouse-derived Serum Albumin/ALB protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-93746 Supplier Medchemexpress LLC Supplier No. HYP78245100UG
Serum albumin/ALB protein binds to various substances including water calcium sodium potassium fatty acids hormones bilirubin and drugs Its main function is to regulate blood osmotic pressure and transport zinc calcium and magnesium Serum Albumin/ALB Protein Mouse (HEK293 His) is the recombinant mouse-derived Serum Albumin/ALB protein expressed by HEK293 with C-His labeled tag