Learn More
Medchemexpress LLC Serum Albumin/ALB M 50ug
SDP
Catalog No. 5000195636
Encompass_Preferred
Recombinant mouse serum albumin produced in Pichia pastoris with an N-terminal 6xHis tag. Supplied as a purified protein (50 μg, ~90% purity, ~67.9 kDa) for use in biochemical assays, antibody production, protein interaction studies, and related research applications. Store at -20°C; shipped on blue ice to preserve integrity.
  • High purity (~90%) for reliable experimental results.
  • Pichia pastoris expression supports eukaryotic folding and modifications.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Compact 50 μg size suited for small-scale assays and pilot studies.
  • Stable when stored at -20°C for extended periods.
  • Shipped on blue ice to maintain the cold chain during transport.
Catalog No. 50-001-95636 Supplier Medchemexpress LLC Supplier No. HYP7171050UG
Recombinant mouse serum albumin produced in Pichia pastoris with an N-terminal 6xHis tag. Supplied as a purified protein (50 μg, ~90% purity, ~67.9 kDa) for use in biochemical assays, antibody production, protein interaction studies, and related research applications. Store at -20°C; shipped on blue ice to preserve integrity.
  • High purity (~90%) for reliable experimental results.
  • Pichia pastoris expression supports eukaryotic folding and modifications.
  • N-terminal 6xHis tag enables affinity purification and detection.
  • Compact 50 μg size suited for small-scale assays and pilot studies.
  • Stable when stored at -20°C for extended periods.
  • Shipped on blue ice to maintain the cold chain during transport.