Learn More
Medchemexpress LLC Serum Albumin/ALB M 5ug
SDP
Catalog No. 5000195917
Encompass_Preferred
Recombinant mouse serum albumin (ALB) expressed in Pichia pastoris with an N-terminal 6xHis tag. Supplied as a lyophilized powder (5 μg) formulated in PBS with trehalose, the protein is >90% pure by reducing SDS-PAGE and has an approximate molecular weight of 67.9 kDa. Store frozen for long-term stability; reconstituted material is stable short term at 4°C.
  • Provides high-purity mouse albumin for biochemical and binding assays.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • Expressed in Pichia pastoris to support eukaryotic post-translational processing.
  • N-terminal His tag enables affinity-based purification or detection.
  • Formulated with trehalose to preserve structural integrity during storage.
  • Validated purity and molecular weight by reducing SDS-PAGE.
Catalog No. 50-001-95917 Supplier Medchemexpress LLC Supplier No. HYP717105UG
Recombinant mouse serum albumin (ALB) expressed in Pichia pastoris with an N-terminal 6xHis tag. Supplied as a lyophilized powder (5 μg) formulated in PBS with trehalose, the protein is >90% pure by reducing SDS-PAGE and has an approximate molecular weight of 67.9 kDa. Store frozen for long-term stability; reconstituted material is stable short term at 4°C.
  • Provides high-purity mouse albumin for biochemical and binding assays.
  • Lyophilized formulation for long-term storage and easy reconstitution.
  • Expressed in Pichia pastoris to support eukaryotic post-translational processing.
  • N-terminal His tag enables affinity-based purification or detection.
  • Formulated with trehalose to preserve structural integrity during storage.
  • Validated purity and molecular weight by reducing SDS-PAGE.