Learn More
Medchemexpress LLC Serum Albumin/ALB R 5ug | 5ug
SDP
Catalog No. 5000194080
Encompass_Preferred
ALB protein is a serum protein synthesized by the liver and plays a vital role in maintaining body fluid balance and transporting various molecules in the blood It is involved in the regulation of osmotic pressure and acts as a carrier for hormones fatty acids and drugs Serum Albumin/ALB Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived Serum Albumin/ALB protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-94080 Supplier Medchemexpress LLC Supplier No. HYP787375UG
ALB protein is a serum protein synthesized by the liver and plays a vital role in maintaining body fluid balance and transporting various molecules in the blood It is involved in the regulation of osmotic pressure and acts as a carrier for hormones fatty acids and drugs Serum Albumin/ALB Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived Serum Albumin/ALB protein expressed by HEK293 with C-His labeled tag