Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Serum albumin, rat (recombinant, HEK293, His tag) | >95.0% | 20UG
SDP

Catalog No. 5000176543
Encompass_Preferred

Serum Albumin Protein, Rat (HEK293, His) is a recombinant rat-derived serum albumin produced in HEK293 cells and engineered with an N-terminal His tag. Supplied as a purified recombinant protein in a small 20 μg format, it is intended for research use in biochemical assays, binding studies, and assay development requiring native-like rat albumin.

  • Recombinant rat serum albumin expressed in HEK293 cells.
  • N-terminal His tag for affinity purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular weight ~67.5 kDa; observed 65-70 kDa.
  • Supplied in a small purified format suitable for assay development.

Catalog No. 50-001-76543 Supplier Medchemexpress LLC Supplier No. HYP70369120UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Serum Albumin Protein, Rat (HEK293, His) is a recombinant rat-derived serum albumin produced in HEK293 cells and engineered with an N-terminal His tag. Supplied as a purified recombinant protein in a small 20 μg format, it is intended for research use in biochemical assays, binding studies, and assay development requiring native-like rat albumin.

  • Recombinant rat serum albumin expressed in HEK293 cells.
  • N-terminal His tag for affinity purification and detection.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular weight ~67.5 kDa; observed 65-70 kDa.
  • Supplied in a small purified format suitable for assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.