Learn More
BioVendor, LLC Serum Amyloid A Human Elisa BioVendor LLC
SDP
Supplier:  BioVendor, LLC RBL008R
Encompass
Serum amyloid A (SAA) is a highly sensitive acute phase reactant that has been linked to inflammatory diseases, both infectious and non-infectious origin. SAA is an apolipoprotein that interacts with HDL, promotes the accumulation of leukocytes at the site of inflammation and the adhesion of platelets, and participates in the removal of damaged cell membranes. The acute phase SAAs (SAA1, SAA2) are transcriptionally regulated in hepatocytes by a variety of inflammatory cytokines, and they can transiently increase > 1000-fold. SAA can be also used to risk prediction of coronary heart disease, the prognosis of tumour patients, and observation of transplant rejection. One kit (96 wells). Type: sandwich ELISA. Sample type: serum, plasma. Sample req: 5 ¿l/well. Cal range: 6.25-200ng/ml. LoD: 1.56ng/ml. Other names: PIG4, SAA1 and SAA2, TP53I4 serum amyloid A1 and serum amyloid A2.
Catalog No. 50-240-6381