Learn More
Cusabio Technology LLC Serum amyloid A1 SAA1
Catalog No. 5000050607
Encompass_Preferred
This kit detects SAA1 in mouse serum plasma and tissue homogenates with a detection range of 3 12 ng/ml to 200 ng/ml and a sensitivity of 0 78 ng/ml Designed for quantitative analysis it employs a sandwich ELISA technique requiring 50-100 l of sample per test Results can be obtained within 1 to 5 hours with measurements taken at 450 nm The assay provides reliable detection of SAA1 protein levels suitable for research applications involving inflammation monitoring or disease biomarker studies in murine models
Catalog No. 50-000-50607 Supplier Cusabio Technology LLC Supplier No. CSBEL020656MO
This kit detects SAA1 in mouse serum plasma and tissue homogenates with a detection range of 3 12 ng/ml to 200 ng/ml and a sensitivity of 0 78 ng/ml Designed for quantitative analysis it employs a sandwich ELISA technique requiring 50-100 l of sample per test Results can be obtained within 1 to 5 hours with measurements taken at 450 nm The assay provides reliable detection of SAA1 protein levels suitable for research applications involving inflammation monitoring or disease biomarker studies in murine models