Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Serum Bottles Clear Wheaton 400, 10 ml, 25 mm diameter, 54 mm height
SDP

Catalog No. 501898936
Encompass_Preferred

Manufactured from Wheaton borosilicate molded glass, meets USP Type I standards.
Volume: 10 ml
Diameter: 25 mm
Height: 54 mm
Material: Wheaton "400"
Cap: 13 x 20 mm

Catalog No. 50-189-8936 Supplier Electron Microscopy Sciences Supplier No. 6030610
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured from Wheaton borosilicate molded glass, meets USP Type I standards.
Volume: 10 ml
Diameter: 25 mm
Height: 54 mm
Material: Wheaton "400"
Cap: 13 x 20 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.