Learn More
Arbor Assays Creatinine Serum Low Sample Volume Kit (384-well Plate)
SDP
Catalog No. 502278529
Encompass_Preferred
The DetectX Low Volume Serum Creatinine kit quantitatively measures creatinine present in serum samples using a 384-well plate format to minimize sample usage.
This item is not returnable. View return policy
Catalog No. 50-227-8529 Supplier Arbor Assays Supplier No. KB02H1D
The DetectX Low Volume Serum Creatinine kit quantitatively measures creatinine present in serum samples using a 384-well plate format to minimize sample usage.