Learn More
BioVendor, LLC SERUM CROSSLAP CTX-I ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC AC02F1
Encompass
TypeSandwich ELISA, HRP-labelled antibodyDescriptionThe Serum CrossLaps (CTX-I) ELISA is an enzyme immunological test for the quantification of degradation products of C-terminal telopeptides of Type I collagen in human serum and plasma.The Serum CrossLaps (CTX-I) ELISA assay is intended for in-vitro diagnostic use as an indication of human bone resorption.ApplicationsSerum, PlasmaSample Requirements50 μl/wellCalibration Range0-3.380 ng/mlLimit of Detection0.020 ng/mL CrossLaps
Catalog No. 50-196-3834