Learn More
STEMCELL Technologies ImmunoCult™-ACF Human B Cell Expansion Supplement, Size: 2 mL
SDP
Catalog No. 502045531
Encompass
ImmunoCult™-ACF Human B Cell Expansion Supplement is a serum-free and animal component-free supplement formulated to promote robust in vitro expansion of human peripheral blood B cells in the absence of serum, feeder cells, or specialized culture plates. The supplement contains recombinant proteins and chemically defined components that support the activation and expansion of human B cells and their maturation to plasma cells.Available either individually or as part of ImmunoCult™ Human B Cell Expansion Kit (/products/immunocult-human-b-cell-expansion-kit.html). ImmunoCult™-ACF Human B Cell Expansion Supplement is a 50X supplement that can be added to a base medium such as ImmunoCult™-XF B Cell Base Medium (/products/immunocult-xf-b-cell-base-medium.html) to prepare a complete serum-free medium for culturing and expanding human B cells.
This item is not returnable. View return policy
Catalog No. 50-204-5531 Supplier STEMCELL Technologies Supplier No. 10974
ImmunoCult™-ACF Human B Cell Expansion Supplement is a serum-free and animal component-free supplement formulated to promote robust in vitro expansion of human peripheral blood B cells in the absence of serum, feeder cells, or specialized culture plates. The supplement contains recombinant proteins and chemically defined components that support the activation and expansion of human B cells and their maturation to plasma cells.Available either individually or as part of ImmunoCult™ Human B Cell Expansion Kit (/products/immunocult-human-b-cell-expansion-kit.html). ImmunoCult™-ACF Human B Cell Expansion Supplement is a 50X supplement that can be added to a base medium such as ImmunoCult™-XF B Cell Base Medium (/products/immunocult-xf-b-cell-base-medium.html) to prepare a complete serum-free medium for culturing and expanding human B cells.