Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Serum Free Cryo Medium (2X), 50mL
SDP

Catalog No. 502209766
Encompass_Preferred

QBIs Serum Free Cryo Medium is a sterile 2X concentrated freezing medium containing DMSO developed for the cryopreservation of cells grown under serum free conditions. Serum Free Cryo Medium is an animal-free, protein-free, chemically defined medium containing DMSO and other cryoprotective components.

With DMSO
Without Phenol Red

Storage Conditions: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Performance Tested
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-220-9766 Supplier Quality Biological Inc Supplier No. 163320051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

QBIs Serum Free Cryo Medium is a sterile 2X concentrated freezing medium containing DMSO developed for the cryopreservation of cells grown under serum free conditions. Serum Free Cryo Medium is an animal-free, protein-free, chemically defined medium containing DMSO and other cryoprotective components.

With DMSO
Without Phenol Red

Storage Conditions: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
Performance Tested
Max Shelf Life: 24 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.