Learn More
STEMCELL Technologies STEMdiff™ Lung Progenitor Kit, Size: 1 Kit
SDP
Catalog No. 502036626
Encompass
STEMdiff™ Lung Progenitor Kit is a serum-free culture medium system for efficient and reproducible generation of lung progenitor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells through three stages of differentiation: 1) definitive endoderm, 2) anterior foregut endoderm, and 3) lung progenitor cells. Differentiated cells will express NKX2.1, a key marker of lung progenitor cells. The resulting cells can be further matured towards proximal or distal airway cells using published protocols for the study of lung diseases and lung development.
Catalog No. 50-203-6626 Supplier STEMCELL Technologies Supplier No. 1000230
STEMdiff™ Lung Progenitor Kit is a serum-free culture medium system for efficient and reproducible generation of lung progenitor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells through three stages of differentiation: 1) definitive endoderm, 2) anterior foregut endoderm, and 3) lung progenitor cells. Differentiated cells will express NKX2.1, a key marker of lung progenitor cells. The resulting cells can be further matured towards proximal or distal airway cells using published protocols for the study of lung diseases and lung development.