Learn More
Cayman Chemical Serum Iron asy KIt 192Wel
SDP
Supplier:  Cayman Chemical 702870192 WELL
Encompass_Preferred
Cayman’s Serum Iron Assay Kit provides a convenient colorimetric method of determining iron levels in serum. This assay has a range of 0-100 µM (0-560 UG/dl) and a lower limit of detection (LLOD) of 0.68 µM (3.8 UG/dl).
Catalog No. 50-289-3982