Learn More
Verichem Laboratories Verichem Serum ISE Standard Kit, 5 x 15.0 mL
Catalog No. NC1168736
Encompass_Preferred
The Verichem Serum ISE Standard Kit from Block Scientific contains 5 bottles of 15 mL each of standard solutions for sodium, potassium, and chloride ion selective electrode calibration. The kit includes sodium standards of 100, 140, and 180 mmol/L; potassium standards of 2, 4, and 8 mmol/L; and chloride standards of 80, 100, and 120 mmol/L. These aqueous, ready-to-use standards allow quick and accurate calibration and quality control checks for ion selective electrode analyzers measuring electrolytes in serum. The standards are traceable to NIST standard reference materials. The kit provides economical, high-quality standards for ensuring accuracy of serum electrolyte measurements.
Catalog No. NC1168736 Supplier Verichem Laboratories Supplier No. 9240
The Verichem Serum ISE Standard Kit from Block Scientific contains 5 bottles of 15 mL each of standard solutions for sodium, potassium, and chloride ion selective electrode calibration. The kit includes sodium standards of 100, 140, and 180 mmol/L; potassium standards of 2, 4, and 8 mmol/L; and chloride standards of 80, 100, and 120 mmol/L. These aqueous, ready-to-use standards allow quick and accurate calibration and quality control checks for ion selective electrode analyzers measuring electrolytes in serum. The standards are traceable to NIST standard reference materials. The kit provides economical, high-quality standards for ensuring accuracy of serum electrolyte measurements.