Description
- Use for discard tubes, immunohematology or viral marker testing
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes have an 18-month shelf life from the date of manufacture
- Nonscrew Safety Pull Caps can be removed with a gentle pull
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Material | Plastic |
| Closure Type | Non-ridged Pull Cap |
| Closure Color | White with Black Ring |
| Tube Size | 13 x 75 mm |