Learn More
Innovative Research Inc Pooled Human Serum Off The Clot 1000ml
Catalog No. 502036418
Encompass_Preferred
Pooled Human Serum from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center. The blood is allowed to clot and is then processed into serum via centrifugation, and is frozen immediately after processing. 
Catalog No. 50-203-6418 Supplier Innovative Research Inc Supplier No. ISER1000ML
Pooled Human Serum from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center. The blood is allowed to clot and is then processed into serum via centrifugation, and is frozen immediately after processing. 