Learn More
Zen Bio Inc SERUM/PLASMA FA KIT 96-WELL
SDP
Supplier:  Zen Bio Inc SFA1
Encompass
Serum/Plasma Fatty Acid Detection Kit. kits to analyze the levels of Non-Esterified Fatty Acid(NEFA) in human and animal serum or plasma. The kits are designed for use in a 96-well format.
Catalog No. NC9088408