Learn More
Sigma Aldrich Fine Chemicals Biosciences Serum PlusTM Medium Supple
Catalog No. 502901264
Encompass
SERUM PLUS(TM) contains a low level of gamma irradiated fetal Bovine Serum (fBS) supplemented with proteins that promote nutrient transport cell attachement and cell growth.
Catalog No. 50-290-1264 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 14008C100ML
SERUM PLUS(TM) contains a low level of gamma irradiated fetal Bovine Serum (fBS) supplemented with proteins that promote nutrient transport cell attachement and cell growth.