Learn More
BioVendor, LLC SERUM PREC CARTILAPS ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC AC08F1
Encompass
Sandwich ELISA, HRP-labelled antibodyThe Serum Pre-Clinical CartiLaps (CTX-II) ELISA detects degradation products of C-terminal telopeptides of type II collagen (CTX-II) in animal serum. The test is intended for research use only, not for use in diagnostic procedures.Disruption of the structural integrity of cartilage is the major histological finding in osteoarthritis and rheumatoid arthritis. Type II collagen is the major organic constituent of cartilage and fragments of type II collagen (CTX-II) are being released into circulation and subsequently secreted into urine following degradation of cartilage. The Serum Pre-Clinical CartiLaps (CTX-II) ELISA has been reported to be useful for rat, mice, rabbit, dog, guinea pig and cynomolgus monkey and horse specimens.ApplicationsSerum, Plasma, Cartilage Explant Culture Supernatant and Synovial FluidSample Requirements25 µL/wellCalibration Range0-300 pg/mLLimit of Detection3.7 pg/mL
Catalog No. 50-196-3839