Learn More
Norgen Biotek Corp SERUM PROTEIN DEPLETION KIT
Catalog No. NC2120717
Encompass_Preferred
ProteoSpin™ Abundant Serum Protein Depletion Kitapid and efficient removal of abundant proteins from serum and plasma samplesProcess 10 samples in 30 minutesAllows for visualization of low abundance proteinsConvenient and affordable spin column protocolGeneric protocol allows for depletion from human and other animal samplesPurification is based on spin column chromatography that uses Norgen’s proprietary Silicon Carbide resin separation matrix
This item is not returnable. View return policy
Catalog No. NC2120717 Supplier Norgen Biotek Corp Supplier No. 17300
ProteoSpin™ Abundant Serum Protein Depletion Kitapid and efficient removal of abundant proteins from serum and plasma samplesProcess 10 samples in 30 minutesAllows for visualization of low abundance proteinsConvenient and affordable spin column protocolGeneric protocol allows for depletion from human and other animal samplesPurification is based on spin column chromatography that uses Norgen’s proprietary Silicon Carbide resin separation matrix