Learn More
MOLECULAR DEPOT LLC Serum Proteins 20 g/dL - 1 mL
Catalog No. 5000013461
Encompass_Preferred
Human Serum Proteins for Interference Testing a highly concentrated solution of human serum proteins (20 g/dL) provided with the corresponding diluent This solution can be spiked into specimens to test human serum proteins interference Instructions with sample calculations are also provided For in vitro research use only
Catalog No. 50-000-13461 Supplier MOLECULAR DEPOT LLC Supplier No. K2010001E
Human Serum Proteins for Interference Testing a highly concentrated solution of human serum proteins (20 g/dL) provided with the corresponding diluent This solution can be spiked into specimens to test human serum proteins interference Instructions with sample calculations are also provided For in vitro research use only