Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Serum, Rabbit, Sterile

Catalog No. MP219135791 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219135791 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219135791 Supplier MP Biomedicals™ Supplier No. 0219135791

missing translation for 'validMainframeMsg'

Serum, Rabbit, Sterile

  • Rabbit serum is a component of blood obtained from a rabbit that contains blood plasma without fibrinogen or other clotting factors.
  • Rabbit Serum Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.
  • In immunohistochemistry, antibodies are used to detect the presence and abundance of certain antigens in a given tissue sample.
  • The use of a blocking serum such as rabbit serum is important because it prevents nonspecific binding and interaction between the antibodies and non-targeted antigens.
  • Rabbit Serum is also a common component in media used to maintain cell culture.
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Content And Storage Store at -20°C.
Form Solution
Product Type Serum Replacement
Quantity 10 x 100 mL
Sterility Sterile
pH 6.5 to 8.0
Protein Content 4.0 to 7.0 gm/dl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.