Learn More
Sigma Aldrich Fine Chemicals Biosciences Serum Replacement 3 50x li
Supplier:  Sigma Aldrich Fine Chemicals Biosciences S2640100ML
Encompass
Sigmas Serum Replacement 3 is a fortified multipurpose serum replacement for the long-term culturing of anchorage dependant and suspension cultures. This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
Catalog No. 50-290-4575