Learn More
Electron Microscopy Sciences Serum Tube Vials Amber, 2 ml
SDP
Catalog No. 501898921
Encompass_Preferred
Manufactured from Wheaton “320" borosilicate glass. Meets ASTM Type I, Class B and USP Type I Standards.
Volume: 2 ml
Catalog No. 50-189-8921 Supplier Electron Microscopy Sciences Supplier No. 6030202
Manufactured from Wheaton “320" borosilicate glass. Meets ASTM Type I, Class B and USP Type I Standards.
Volume: 2 ml