Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™Serum Toxicology Benzodiazepine Assay

Catalog No. 23278327 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 x 8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-327 25 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-327 Supplier Thermo Scientific™ Supplier No. 0920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utilize this serum toxicology Benzodiazepine assay in emergency room situations where urine samples are not conveniently available.

Utilize Thermo Scientific™ DRI™ Serum Toxicology Benzodiazepine Assay in emergency room situations where urine samples are not conveniently available.

  • Serum-based assay
  • Used to indentify class of drug of overdose
  • Conveniently packaged with separate calibrators for ease of use
  • Liquid, ready-to-use and has the flexibility to run on a variety of clinical chemistry analyzers

Ordering Alerts:

0920: Reagent volume; 25, 8mL

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Serum Toxicology Benzodiazepine
Detectable Analytes Benzodiazepines (BZO)
Sample Type Serum
Type Reagent
Quantity 25 x 8 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.