Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Serum Toxicology Calibrator 1

Catalog No. 23278329 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-329 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-329 Supplier Thermo Scientific™ Supplier No. 0963
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use with serum toxicology immunoassays.

Use with the serum toxicology Barbiturate, serum toxicology Benzodiazepine, and Tricyclics immunoassays. The Thermo Scientific™ DRI™ Serum Toxicology Calibrator 1 is a liquid, ready-to-use calibrators conveniently packaged for use with the serum toxicology immunoassays.

  • Multianalyte calibrators

Ordering Information:

0963: 1 bottle, 5mL. Concentrations: Barbiturate 500ng/mL, Benzodiazepine 25ng/mL, Tricyclics 150ng/mL.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Serum Toxicology Calibrator 1
Detectable Analytes Benzodiazepines, Tricyclic Antidepressants, Serum Barbiturate, Serum Benzodiazepine, Serum Tricyclic
Sample Type Serum
Type Calibrator
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.