Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Serum Toxicology Calibrator 2

Catalog No. 23278330 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-330 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-330 Supplier Thermo Scientific™ Supplier No. 0965
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Thermo Scientific™ DRI™ Serum Toxicology Calibrator 2 with Serum Toxicology Barbiturate, Serum Toxicology Benzodiazepine, and Tricyclics Immunoassays.

Use Thermo Scientific™ DRI™ Serum Toxicology Calibrator 2 with Serum Toxicology Barbiturate, Serum Toxicology Benzodiazepine and Tricyclics Immunoassays. Liquid, ready-to-use calibrators are conveniently packaged for use with serum toxicology immunoassays.

  • Multianalyte calibrators

Ordering Alerts:

0965: 1 bottle, 5mL. Concentrations: Barbiturate 1000ng/mL, Benzodiazepine 50ng/mL and Tricyclics 300ng/mL.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Serum Toxicology Calibrator 2
Detectable Analytes Benzodiazepines, Tricyclic Antidepressants, Serum Barbiturate, Serum Benzodiazepine, Serum Tricyclic
Sample Type Serum
Type Calibrator
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.