Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™Serum Toxicology Negative Calibrator

Catalog No. 23278328 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-328 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-328 Supplier Thermo Scientific™ Supplier No. 0962
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure accurate results in barbiturate, benzodiazepine and tricyclics immunoassays with the Thermo Scientific™ DRI™ Serum Toxicology Negative Calibrator.

Ensure accurate results in barbiturate, benzodiazepine and tricyclics immunoassays with the Thermo Scientific™ DRI™ Serum Toxicology Negative Calibrator. Liquid, ready-to-use calibrators are conveniently packaged for use with the serum toxicology immunoassays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Serum Toxicology Negative Calibrator
Detectable Analytes Negative Calibrator
Sample Type Serum
Type Calibrator
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.