Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One VACUETTE™ CAT Serum Separator Clot Activator Blood Collection Tubes

Catalog No. 22040547
Change view
Click to view available options
Capacity (Metric):
2.5 mL
3.5 mL
4 mL
5 mL
7 mL
8 mL
9 mL
Closure Color:
Gold with Gold Ring
Red with White Ring
Red with Yellow Ring
Tube Size:
13 x 75 mm
13 x 100 mm
16 x 100 mm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color Tube Size
22-040-547 5 mL Gold with Gold Ring 13 x 100 mm
22-040-542 2.5 mL Red with White Ring 13 x 75 mm
22-040-536 3.5 mL Red with Yellow Ring 13 x 75 mm
22-040-545 4 mL Red with Yellow Ring 13 x 100 mm
22-040-541 5 mL Red with Yellow Ring 13 x 100 mm
22-040-538 7 mL Red with Yellow Ring 16 x 100 mm
22-040-546 3.5 mL Gold with Gold Ring 13 x 75 mm
22-040-537 8 mL Red with Yellow Ring 16 x 100 mm
22-040-544 9 mL Red with Yellow Ring 16 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-040-547 Supplier Greiner Bio-One Supplier No. 456018P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VACUETTE™ CAT Serum Separator Clot Activator tubes are evacuated blood collection tubes used for serum determinations. They contain clot-activating micronized silica particles and a gel separator that, once centrifuged, forms a stable barrier separating the serum from fibrin and cells.

  • VACUETTE™ Blood Collection Tubes are evacuated for easy, safe sample collection. The amount of vacuum in each tube corresponds to the tube’s blood volume specification and helps ensure the correct blood-to-additive ratio.
  • Injection molded from polyethylene terephthalate (PET) plastic, VACUETTE™ Blood Collection Tubes are highly transparent and virtually unbreakable.
  • VACUETTE™ Serum tubes include a clot-activating additive – micronized silica particles – that is spray-dried on the interior tube wall for even disbursement and optimal blood/additive mixing.
  • Serum tubes with gel contain a barrier gel to separate the serum from the fibrin and cells. During centrifugation, the barrier gel moves upward to the serum/clot interface, where it forms a stable barrier. This barrier separates the serum from fibrin and cells.
  • Optimal fill marks on every tube label indicate the proper draw volume for correct blood-to-additive ratios (within ±10% of the stated nominal volume in accordance with ISO and CLSI).
  • Featuring red VACUETTE™ SAFETY Caps, designed to minimize the spraying of aerosols during decapping and color-coded to aid first-glance tube recognition.
  • Sterile and latex-free.
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Product Type Blood Collection Tube
Additive Clot Activator and Gel
Material PET
Sterility Sterile
Closure Type Non-ridged Pull Cap
Capacity (Metric) 5 mL
Diameter (Metric) 13 mm
Closure Color Gold with Gold Ring
Tube Size 13 x 100 mm
Length (Metric) 100 mm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.