Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Serum Vial Racks

Catalog No. 03411762 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Holds:
Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
03-411-762 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-411-762 Supplier Bel-Art™ Supplier No. F185130072
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to withstand elevated water temperatures for incubation.

  • For use with serum vials (10-13 mm dia.), RIA vials and T3/T4 reaction vials (13-16 mm and 16-20 mm dia.)
  • Long-lasting and corrosion-resistant
  • With end plates handles
  • Made from polypropylene
  • Steam autoclavable at 121°C (250°F)

Specifications

Description Corrosion resistant
Autoclavable Autoclavable
Color White
Holds Vials
Material Polypropylene
Positions 72
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) For use with serum vials
Size L x W x H: 23.8 x 11.1 x 8.9 cm
Dimensions (L x W x H) 23.8 x 11.1 x 8.9cm
Length (Metric) 241 mm
Includes End plate handles
Product Type Vial Rack
Diameter (Metric) Inner 13 mm
Shelves 2 Tiers
Height (Metric) 89 mm
Array 12 x 6
Width (Metric) 102 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.