Learn More
ACROBiosystems ACROBiosystems Cynomolgus Serum Albumin Protein, His Tag
Catalog No. 501956446
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus Serum Albumin, His Tag (CSA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ala 608 (Accession # A2V9Z4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.9 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-6446 Supplier ACROBiosystems Supplier No. CSAC52H4100UG
Recombinant Protein;100UG;Cynomolgus Serum Albumin, His Tag (CSA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Ala 608 (Accession # A2V9Z4-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.9 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE).