Learn More
ACROBiosystems ACROBiosystems Mouse Serum Albumin Protein, His Tag
Catalog No. 501956454
Encompass_Preferred
Recombinant Protein;100UG;Mouse Serum Albumin, His Tag (MSA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.8 kDa. The protein migrates as 65-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6454 Supplier ACROBiosystems Supplier No. MSAM52H8100UG
Recombinant Protein;100UG;Mouse Serum Albumin, His Tag (MSA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 67.8 kDa. The protein migrates as 65-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.