Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Serum Albumin Protein, His,Avitag™
Catalog No. 501956694
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Serum Albumin, His,Avitag (HSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Leu 609 (Accession # P02768-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 70.2 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6694 Supplier ACROBiosystems Supplier No. HSAH82E325UG
Recombinant Protein;25UG;Biotinylated Human Serum Albumin, His,Avitag (HSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 25 - Leu 609 (Accession # P02768-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 70.2 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.