Learn More
ATCC Serum-Free Cell Freezing Medium, 1X, 20 ML
Catalog No. 502384460
Encompass
ATCC Serum-Free Cell Freezing Medium is a sterile, ready-to-use medium suitable for the cryopreservation of adherent and suspension cell cultures. This proprietary, serum-free formulation contains 10% DMSO and methylcellulose. Cells cryopreserved using Serum-Free Freezing Medium show levels of viability and percent attachment (adherent cells) that are comparable to cells preserved in DMSO and FBS. Serum-Free Cell Freezing Medium can be used for both cells cultured in serum-supplemented growth medium as well as cells grown under serum-free conditions. This product has applications for cyropreservation, cell culture, cell growth, and viability.
Catalog No. 50-238-4460 Supplier ATCC Supplier No. 302600
ATCC Serum-Free Cell Freezing Medium is a sterile, ready-to-use medium suitable for the cryopreservation of adherent and suspension cell cultures. This proprietary, serum-free formulation contains 10% DMSO and methylcellulose. Cells cryopreserved using Serum-Free Freezing Medium show levels of viability and percent attachment (adherent cells) that are comparable to cells preserved in DMSO and FBS. Serum-Free Cell Freezing Medium can be used for both cells cultured in serum-supplemented growth medium as well as cells grown under serum-free conditions. This product has applications for cyropreservation, cell culture, cell growth, and viability.