Learn More
Crescent Chemical Co Inc SERVA BLUE G
Supplier: Crescent Chemical Co Inc SE35050.02
SERVA Blue G For preparation of staining solution for protein detection in gel electrophoresis. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35050.02 CAS 6104-58-1