Learn More
Crescent Chemical Co Inc SERVA BLUE G SOLN FOR BN 1%
SDP
Catalog No. 50248601
Encompass_Preferred
SERVA Blue G solution for BN, 1% Solution for preparing the blue stained cathode running buffer for Blue Native gel electrophoresis. SERVA Blue G (cat. no. 35050): 10 g/l in aqua dest. Size - 20ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 42538.01 /
Catalog No. 50-248-601 Supplier Crescent Chemical Co Inc Supplier No. SE42538.01
SERVA Blue G solution for BN, 1% Solution for preparing the blue stained cathode running buffer for Blue Native gel electrophoresis. SERVA Blue G (cat. no. 35050): 10 g/l in aqua dest. Size - 20ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 42538.01 /