Learn More
Crescent Chemical Co Inc SERVA BLUE R
SDP
Catalog No. 50248121
Encompass_Preferred
SERVA Blue R For preparation of staining solution for protein detection in gel electrophoresis. R stands for "reddish". Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35051.02 CAS 6104-59-2
Catalog No. 50-248-121 Supplier Crescent Chemical Co Inc Supplier No. SE35051.02
SERVA Blue R For preparation of staining solution for protein detection in gel electrophoresis. R stands for "reddish". Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35051.02 CAS 6104-59-2