Learn More
Crescent Chemical Co Inc SERVA BLUE R
Supplier: Crescent Chemical Co Inc SE35051.03
SERVA Blue R For preparation of staining solution for protein detection in gel electrophoresis. R stands for "reddish". Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35051.03 CAS 6104-59-2