Learn More
Crescent Chemical Co Inc SERVA BLUE W SOLUBLE IN WATER
Supplier: Crescent Chemical Co Inc SE35053.03
SERVA Blue W soluble in water A triphenylmethane dye, more lipophilic methyl homologue of SERVA Violet R. Suitable for round gels of PAGE, also for flat gels 0.1 - 1.0 mm thick. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35