Learn More
Crescent Chemical Co Inc SERVA BLUE W SOLUBLE IN WATER
SDP
Catalog No. 50248123
Encompass_Preferred
SERVA Blue W soluble in water A triphenylmethane dye, more lipophilic methyl homologue of SERVA Violet R. Suitable for round gels of PAGE, also for flat gels 0.1 - 1.0 mm thick. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 350
Catalog No. 50-248-123 Supplier Crescent Chemical Co Inc Supplier No. SE35053.02
SERVA Blue W soluble in water A triphenylmethane dye, more lipophilic methyl homologue of SERVA Violet R. Suitable for round gels of PAGE, also for flat gels 0.1 - 1.0 mm thick. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 350